6-Aminonicotinonitrile, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Amino-3-pyridinecarbonitrile, 6-aminopyridine-3-carbonitrile, 2-Amino-5-cyanopyridine, 2-Aminopyridine-5-carbonitrile
M-Clarity™ quality level = MQ100
637475-25G
637475-5G
SIAL637475-25GEA
3210
CZK
SIAL637475-25G
SIAL637475-5G
6-Aminonicotinonitrile, Sigma-Aldrich®
6-Aminonicotinonitrile
Vzorec:
C₆H₅N₃ MW: 119,13 g/mol Bod varu: 240…250 °C (15 mmHg) Bod tání: 164 °C |
MDL Number:
MFCD00128902 CAS číslo: 4214-73-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...