Tisk…

5-Chloro-2-thenoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Chlorothiophene-2-carboxylic acid, 5-Chloro-2-thiophenecarboxylic acid

M-Clarity™ quality level = MQ200

633003-25G 633003-5G
SIAL633003-25GEA 4650 CZK
SIAL633003-25G SIAL633003-5G
5-Chloro-2-thenoic acid, Sigma-Aldrich®
5-Chloro-2-thenoic acid
Vzorec: C₅H₃ClO₂S
MW: 162,6 g/mol
MDL Number: MFCD00041426
CAS číslo: 24065-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR