Tisk…

Indole-4-carbaldehyd, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1H-Indole-4-carbaldehyde, Indole-4-carboxaldehyde

M-Clarity™ quality level = MQ100

632422-1G 632422-5G
SIAL632422-1GEA 11200 CZK
SIAL632422-1G SIAL632422-5G
Indole-4-carbaldehyd, Sigma-Aldrich®
Indole-4-carbaldehyd
Vzorec: C₉H₇NO
MW: 145,16 g/mol
MDL Number: MFCD01632221
CAS číslo: 1074-86-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR