3-Fluoro-N-methylaniline, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
630969-1G
630969-5G
SIAL630969-1GEA
0
CZK
SIAL630969-1G
SIAL630969-5G
3-Fluoro-N-methylaniline, Sigma-Aldrich®
3-Fluoro-N-methylaniline
Vzorec:
C₇H₈FN MW: 125.15 |
MDL Number:
MFCD02683095 CAS číslo: 1978-37-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...