Tisk…

3,5-Di-tert-butylbenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,5-Bis(tert-butyl)benzaldehyde

M-Clarity™ quality level = MQ200

574589-1G 574589-5G
SIAL574589-1GEA 2890 CZK
SIAL574589-1G SIAL574589-5G
3,5-Di-tert-butylbenzaldehyde, Sigma-Aldrich®
3,5-Di-tert-butylbenzaldehyde
Vzorec: C₁₅H₂₂O
MW: 218,34 g/mol
MDL Number: MFCD00026298
CAS číslo: 17610-00-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR