Tisk…

6-Aminobenzothiazole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Amino-1,3-benzothiazole, Benzothiazol-6-ylamine

M-Clarity™ quality level = MQ100

562440-1G 562440-5G
SIAL562440-1GEA 1460 CZK
SIAL562440-1G SIAL562440-5G
6-Aminobenzothiazole, Sigma-Aldrich®
6-Aminobenzothiazole
Vzorec: C₇H₆N₂S
MDL Number: MFCD00015461
CAS číslo: 533-30-2
EINECS: 208-559-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR