4-Benzylaniline, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Aminodiphenylmethane
M-Clarity™ quality level = MQ100
560235-1G
560235-5G
SIAL560235-1GEA
1760
CZK
SIAL560235-1G
SIAL560235-5G
4-Benzylaniline, Sigma-Aldrich®
4-Benzylaniline
Vzorec:
C₁₃H₁₃N MW: 183,25 g/mol Bod varu: 300 °C (1013 hPa) Bod tání: 35…36 °C Hustota: 1,07 g/cm³ (20 °C) |
MDL Number:
MFCD00047861 CAS číslo: 1135-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...