Tisk…

1-Ethyl-3-methylimidazolium ethylsulphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: EMIM ESU

51682-1KG-F 51682-100G-F
SIAL51682-1KG-FEA 12550 CZK
SIAL51682-1KG-F SIAL51682-100G-F
1-Ethyl-3-methylimidazolium ethylsulphate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium ethylsulphate
Vzorec: C₈H₁₆N₂O₄S
MW: 236,29 g/mol
MDL Number: MFCD06798189
CAS číslo: 342573-75-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR