6-Chloro-1-hexyne, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Chlorohex-1-yne
M-Clarity™ quality level = MQ100
469777-25ML
469777-5ML
SIAL469777-25MLEA
8530
CZK
SIAL469777-25ML
SIAL469777-5ML
6-Chloro-1-hexyne, Sigma-Aldrich®
6-Chloro-1-hexyne
Vzorec:
C₆H₉Cl MW: 116,59 g/mol |
MDL Number:
MFCD00013697 CAS číslo: 10297-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...