Tisk…

3-Fluorophthalic anhydride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Fluoroisobenzofuran-1,3-dione

M-Clarity™ quality level = MQ200

467707-25G 467707-5G
SIAL467707-25GEA 16110 CZK
SIAL467707-25G SIAL467707-5G
3-Fluorophthalic anhydride, Sigma-Aldrich®
3-Fluorophthalic anhydride
Vzorec: C₈H₃FO₃
MW: 166,11 g/mol
Bod varu: 351 °C (1013 hPa)
Bod tání: 162…163 °C
MDL Number: MFCD00039696
CAS číslo: 652-39-1
EINECS: 211-491-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR