4-Fluoro-2-(trifluoromethyl)benzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
436097-1G
436097-5G
SIAL436097-1GEA
0
CZK
SIAL436097-1G
SIAL436097-5G
4-Fluoro-2-(trifluoromethyl)benzaldehyde, Sigma-Aldrich®
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₄F₄O MW: 192,11 g/mol Bod varu: 114 °C (1013 hPa) Hustota: 1,4 g/cm³ (20 °C) |
MDL Number:
MFCD00061313 CAS číslo: 90176-80-0 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...