Tisk…

3-(o-Tolyl)propionic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-(2-Methylphenyl)propionic acid, 2-Methylhydrocinnamic acid, 3-(2-Methylphenyl)propanoic acid

M-Clarity™ quality level = MQ100

433330-10G 433330-1G
SIAL433330-10GEA 5860 CZK
SIAL433330-10G SIAL433330-1G
3-(o-Tolyl)propionic acid, Sigma-Aldrich®
3-(o-Tolyl)propionic acid
Vzorec: C₁₀H₁₂O₂
MDL Number: MFCD00079773
CAS číslo: 22084-89-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR