Tisk…

5-Bromo-1-indanone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Bromo-2,3-dihydro-1H-inden-1-one

M-Clarity™ quality level = MQ200

433098-1G 433098-5G
SIAL433098-1GEA 1320 CZK
SIAL433098-1G SIAL433098-5G
5-Bromo-1-indanone, Sigma-Aldrich®
5-Bromo-1-indanone
Vzorec: C₉H₇BrO
MW: 211,06 g/mol
Bod varu: 280 °C (1013 hPa)
Bod tání: 126…127 °C
MDL Number: MFCD00082718
CAS číslo: 34598-49-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR