5-Bromo-2,4-dichloropyrimidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
416762-25G
416762-5G
SIAL416762-25GEA
11400
CZK
SIAL416762-25G
SIAL416762-5G
5-Bromo-2,4-dichloropyrimidine, Sigma-Aldrich®
5-Bromo-2,4-dichloropyrimidine
Vzorec:
C₄HBrCl₂N₂ MW: 227,87 g/mol Bod varu: 288 °C (1013 hPa) Bod tání: –1 °C Hustota: 1,781 g/cm³ (20 °C) |
MDL Number:
MFCD00127818 CAS číslo: 36082-50-5 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...