1-(2-Pyridyl)piperazine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(1-Piperazinyl)pyridine
M-Clarity™ quality level = MQ100
408166-25ML
408166-5ML
SIAL408166-25MLEA
6050
CZK
SIAL408166-25ML
SIAL408166-5ML
1-(2-Pyridyl)piperazine, Sigma-Aldrich®
1-(2-Pyridyl)piperazine
Vzorec:
C₉H₁₃N₃ Bod varu: 293 °C (1013 hPa) Bod tání: 85…87 °C Hustota: 1,07 g/cm³ (20 °C) |
MDL Number:
MFCD00006216 CAS číslo: 34803-66-2 EINECS: 252-220-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...