Tisk…

Tin(II) oxalate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Tin (II) ethanedioate, Oxalic acid tin (II) salt

M-Clarity™ quality level = MQ100

402761-100G 402761-500G
SIAL402761-100GEA 1380 CZK
SIAL402761-100G SIAL402761-500G
Tin(II) oxalate, Sigma-Aldrich®
Tin(II) oxalate
Vzorec: C₂O₄Sn
Hustota: 3,55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040678
CAS číslo: 814-94-8
EINECS: 212-414-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,08865

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR