N-Ethylisopropylamine, Sigma-Aldrich®
Dodavatel: Merck
MQ200
386391-25ML
386391-5ML
SIAL386391-25MLEA
9110
CZK
SIAL386391-25ML
SIAL386391-5ML
N-Ethylisopropylamine, Sigma-Aldrich®
N-Ethylisopropylamine
Vzorec:
C₅H₁₃N MW: 87.16 Bod varu: 60…62 °C (1013 hPa) Hustota: 0.7085 g/cm³ (20 °C) |
MDL Number:
MFCD00015105 CAS číslo: 19961-27-4 EINECS: 243-442-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...