Sodium perborate monohydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
372862-100G
372862-500G
SIAL372862-100GEA
3620
CZK
SIAL372862-100G
SIAL372862-500G
Sodium perborate monohydrate, Sigma-Aldrich®
Sodium perborate monohydrate
Vzorec:
BH₂NaO₄·1H₂O MW: 117.83 Storage Temperature: Ambient |
MDL Number:
MFCD00149230 CAS číslo: 10332-33-9 EINECS: 239-172-9 UN: 3377 ADR: 5.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...