Tisk…

cis-1,2-Cyclopentanediol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

361437-1G 361437-5G
SIAL361437-1GEA 1820 CZK
SIAL361437-1G SIAL361437-5G
cis-1,2-Cyclopentanediol, Sigma-Aldrich®
cis-1,2-Cyclopentanediol
Vzorec: C₅H₁₀O₂
MW: 102.13
Bod varu: 229
Bod tání: 30
MDL Number: MFCD00134192
CAS číslo: 5057-98-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR