Sodium dichloroacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Sodium 2,2-dichloroacetate
M-Clarity™ quality level = MQ100
347795-10G
347795-50G
SIAL347795-10GEA
2710
CZK
SIAL347795-10G
SIAL347795-50G
Sodium dichloroacetate, Sigma-Aldrich®
Sodium dichloroacetate
Vzorec:
C₂HCl₂NaO₂ MW: 150,92 g/mol Bod tání: 198…199 °C |
MDL Number:
MFCD00070489 CAS číslo: 2156-56-1 EINECS: 218-461-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...