Tisk…

2-Phenoxyaniline, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Amino-2-phenoxybenzol

M-Clarity™ quality level = MQ100

346683-25G 346683-500G
SIAL346683-25GEA 1230 CZK
SIAL346683-25G SIAL346683-500G
2-Phenoxyaniline, Sigma-Aldrich®
2-Phenoxyaniline
Vzorec: C₁₂H₁₁NO
MW: 185,23 g/mol
Bod varu: 170 °C (18 mmHg)
Bod tání: 46…48 °C
MDL Number: MFCD00035765
CAS číslo: 2688-84-8
EINECS: 220-254-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR