Tisk…

N-Methylethylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Ethylmethylamine

M-Clarity™ quality level = MQ200

291145-25G 291145-5G
SIAL291145-25GEA 7770 CZK
SIAL291145-25G SIAL291145-5G
N-Methylethylamine, Sigma-Aldrich®
N-Methylethylamine
Vzorec: C₃H₉N
MW: 59.11
Bod varu: 36…37 °C (1013 hPa)
Hustota: 0.6893
MDL Number: MFCD00009030
CAS číslo: 624-78-2
EINECS: 210-862-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR