Tisk…

2,4-Bis(trifluoromethyl)benzyl bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(Bromomethyl)-2,4-bis(trifluoromethyl)benzene

M-Clarity™ quality level = MQ100

290149-1G 290149-5G
SIAL290149-1GEA 1750 CZK
SIAL290149-1G SIAL290149-5G
2,4-Bis(trifluoromethyl)benzyl bromide, Sigma-Aldrich®
2,4-Bis(trifluoromethyl)benzyl bromide
Vzorec: C₉H₅BrF₆
Bod varu: 112 °C (0,2 mmHg)
Hustota: 1,637 g/cm³ (20 °C)
MDL Number: MFCD00010306
CAS číslo: 140690-56-8
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR