Diethyl methylphosphonate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
268119-25G
268119-5G
SIAL268119-25GEA
10360
CZK
SIAL268119-25G
SIAL268119-5G
Diethyl methylphosphonate, Sigma-Aldrich®
Diethyl methylphosphonate
Vzorec:
C₅H₁₃O₃P MW: 152.13 Bod varu: 190…194 °C (1013 hPa) Hustota: 1.0406 g/cm³ (30 °C) |
MDL Number:
MFCD00009813 CAS číslo: 683-08-9 EINECS: 211-667-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...