4-(Trifluoromethyl)benzylamine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
263508-10G
263508-1G
SIAL263508-10GEA
5580
CZK
SIAL263508-10G
SIAL263508-1G
4-(Trifluoromethyl)benzylamine, Sigma-Aldrich®
4-(Trifluoromethyl)benzylamine
Vzorec:
C₈H₈F₃N Bod varu: 95…97 °C (1013 hPa) Hustota: 1,229 g/cm³ (20 °C) |
MDL Number:
MFCD00010220 CAS číslo: 3300-51-4 EINECS: 221-971-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...