Tisk…

3,5-Bis(trifluoromethyl)benzyl bromide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

263397-1G 263397-5G
SIAL263397-1GEA 1330 CZK
SIAL263397-1G SIAL263397-5G
3,5-Bis(trifluoromethyl)benzyl bromide, Sigma-Aldrich®
3,5-Bis(trifluoromethyl)benzyl bromide
Vzorec: C₉H₅BrF₆
Bod varu: 136…140 °C (14 mmHg)
Hustota: 1,67 g/cm³ (20 °C)
MDL Number: MFCD00009905
CAS číslo: 32247-96-4
EINECS: 250-971-1
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR