3,5-Bis(trifluoromethyl)benzylamine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(3,5-Bis(trifluoromethyl)phenyl)methanamine
M-Clarity™ quality level = MQ100
263389-1G
263389-5G
SIAL263389-1GEA
1810
CZK
SIAL263389-1G
SIAL263389-5G
3,5-Bis(trifluoromethyl)benzylamine, Sigma-Aldrich®
3,5-Bis(trifluoromethyl)benzylamine
Vzorec:
C₉H₇F₆N MW: 243,15 g/mol Bod varu: 82…84 °C (15 mmHg) Bod tání: 46…55 °C |
MDL Number:
MFCD00009909 CAS číslo: 85068-29-7 EINECS: 285-290-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...