Tisk…

8-Bromocaprylic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

257583-10G 257583-1G 257583-50G
SIAL257583-10GEA 6140 CZK
SIAL257583-10G SIAL257583-1G SIAL257583-50G
8-Bromocaprylic acid, Sigma-Aldrich®
8-Bromocaprylic acid
Vzorec: BrH₂C(CH₂)₆COOH
Bod varu: 316 °C (1013 hPa)
Hustota: 1,324 g/cm³ (20 °C)
MDL Number: MFCD00004430
CAS číslo: 17696-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR