Tisk…

Tris(diethylamino)phosphine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

253189-25G 253189-5G
SIAL253189-25GEA 7220 CZK
SIAL253189-25G SIAL253189-5G
Tris(diethylamino)phosphine, Sigma-Aldrich®
Tris(diethylamino)phosphine
Vzorec: C₁₂H₃₀N₃P
Hustota: 0,89 g/cm³ (20 °C)
MDL Number: MFCD00009041
CAS číslo: 2283-11-6
EINECS: 218-920-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR