Tisk…

2-Acetyl-5-chlorothiophene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(5-Chloro-2-thienyl)-1-ethanone, Rivaroxaban Impurity 20

M-Clarity™ quality level = MQ100

247073-25G 247073-5G
SIAL247073-25GEA 0 CZK
SIAL247073-25G SIAL247073-5G
2-Acetyl-5-chlorothiophene, Sigma-Aldrich®
2-Acetyl-5-chlorothiophene
Vzorec: C₆H₅ClOS
MW: 160,62 g/mol
Bod varu: 117 °C (17 mmHg)
Bod tání: 52 °C
MDL Number: MFCD00005444
CAS číslo: 6310-09-4
EINECS: 228-630-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR