Tetrakis(triphenylphosphine)platinum(0), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tetrakis(triphenylphosphine)platinum
M-Clarity™ quality level = MQ100
244961-1G
244961-5G
SIAL244961-1GEA
4730
CZK
SIAL244961-1G
SIAL244961-5G
Tetrakis(triphenylphosphine)platinum(0), Sigma-Aldrich®
Tetrakis(triphenylphosphine)platinum(0)
Vzorec:
C₇₂H₆₀P₄Pt MW: 1244,24 g/mol |
MDL Number:
MFCD00010014 CAS číslo: 14221-02-4 EINECS: 238-087-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...