Allyl isocyanate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Isocyanatoprop-1-ene
M-Clarity™ quality level = MQ100
243272-25G
243272-5G
SIAL243272-25GEA
10990
CZK
SIAL243272-25G
SIAL243272-5G
Allyl isocyanate, Sigma-Aldrich®
Allyl isocyanate
Vzorec:
C₄H₅NO Bod varu: 87…89 °C (1013 hPa) Hustota: 0,94 g/cm³ (20 °C) |
MDL Number:
MFCD00002044 CAS číslo: 1476-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...