Tisk…

5-tert-Butyl-m-xylene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-tert-Butyl-3,5-dimethylbenzene, 1-(tert-Butyl)-3,5-dimethylbenzene

M-Clarity™ quality level = MQ100

233587-100G 233587-500G
SIAL233587-100GEA 1230 CZK
SIAL233587-100G SIAL233587-500G
5-tert-Butyl-m-xylene, Sigma-Aldrich®
5-tert-Butyl-m-xylene
Vzorec: C₁₂H₁₈
MW: 162.27
Bod varu: 205
Bod tání: –22 °C
Hustota: 0.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008832
CAS číslo: 98-19-1
EINECS: 202-647-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR