3-Formylbenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
232793-1G
232793-5G
SIAL232793-1GEA
2050
CZK
SIAL232793-1G
SIAL232793-5G
3-Formylbenzoic acid, Sigma-Aldrich®
3-Formylbenzoic acid
Vzorec:
C₈H₆O₃ Bod varu: 306 °C (1013 hPa) Bod tání: 175 °C |
MDL Number:
MFCD00039575 CAS číslo: 619-21-6 EINECS: 210-585-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...