Trichloromethyl chloroformate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Chloroformic acid trichloromethyl ester, Diphosgene
23261-10ML
23261-50ML
SIAL23261-10MLEA
1680
CZK
SIAL23261-10ML
SIAL23261-50ML
Trichloromethyl chloroformate, Sigma-Aldrich®
Trichloromethyl chloroformate
Vzorec:
C₂Cl₄O₂ MW: 197.83 Bod varu: 128 Bod tání: –57 °C Hustota: 1.63 g/cm³ (20 °C) |
MDL Number:
MFCD00015553 CAS číslo: 503-38-8 EINECS: 207-965-9 Merck Index: 13,03368 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...