N-(Chloromethyl)phthalimide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(Chloromethyl)isoindoline-1,3-dione
M-Clarity™ quality level = MQ200
232424-10G
232424-50G
SIAL232424-10GEA
1630
CZK
SIAL232424-10G
SIAL232424-50G
N-(Chloromethyl)phthalimide, Sigma-Aldrich®
N-(Chloromethyl)phthalimide
Vzorec:
C₉H₆ClNO₂ Bod varu: 305 °C (1013 hPa) Bod tání: 123…126 °C |
MDL Number:
MFCD00005898 CAS číslo: 17564-64-6 EINECS: 241-541-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...