Tisk…

Tetrabutylammonium tetrahydroborate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

230170-10G 230170-50G
SIAL230170-10GEA 3280 CZK
SIAL230170-10G SIAL230170-50G
Tetrabutylammonium tetrahydroborate, Sigma-Aldrich®
Tetrabutylammonium tetrahydroborate
Vzorec: C₁₆H₄₀BN
Bod tání: 124…126 °C
MDL Number: MFCD00012035
CAS číslo: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR