N,N-Dimethyl-1,1,1-trimethylsilylamine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(Dimethylamino)trimethylsilane, Trimethylsilyldimethylamine, N,N-Dimethyltrimethylsilylamine, TMSDMA, N-(Trimethylsilyl)dimethylamine, N,N-Dimethylaminotrimethylsilane, Pentamethylsilylamine
M-Clarity™ quality level = MQ200
226289-10G
226289-50G
SIAL226289-10GEA
1290
CZK
SIAL226289-10G
SIAL226289-50G
N,N-Dimethyl-1,1,1-trimethylsilylamine, Sigma-Aldrich®
N,N-Dimethyl-1,1,1-trimethylsilylamine
Vzorec:
C₅H₁₅NSi Bod varu: 84 °C (1013 hPa) Hustota: 0,7466 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008297 CAS číslo: 2083-91-2 EINECS: 218-222-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...