Tisk…

Jantaran dvojsodný, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,4-Butanedioic acid disodium salt, Sodium Succinate, Succinic acid disodium salt, Sodium succinate dibasic

M-Clarity™ quality level = MQ100

224731-100G 224731-500G 224731-5G
SIAL224731-100GEA 1210 CZK
SIAL224731-100G SIAL224731-500G SIAL224731-5G
Jantaran dvojsodný, Sigma-Aldrich®
Jantaran dvojsodný
Vzorec: C₄H₄Na₂O₄
Hustota: 1,88 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002790
CAS číslo: 150-90-3
EINECS: 205-778-7
Merck Index: 12,08828

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR