Tisk…

Vinyltrimethylsilane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Trimethyl(vinyl)silane

M-Clarity™ quality level = MQ100

213950-100G 213950-25G 213950-5G
SIAL213950-100GEA 40160 CZK
SIAL213950-100G SIAL213950-25G SIAL213950-5G
Vinyltrimethylsilane, Sigma-Aldrich®
Vinyltrimethylsilane
Vzorec: C₅H₁₂Si
MW: 100.24
Bod varu: 55
Hustota: 0.7038 g/cm³ (20 °C)
MDL Number: MFCD00008606
CAS číslo: 754-05-2
EINECS: 212-042-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR