Diammonium tetrachloroplatinate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Ammonium tetrachloroplatinate(II), Ammonium tetrachloroplatinate
M-Clarity™ quality level = MQ100
206105-1G
206105-5G
SIAL206105-1GEA
5370
CZK
SIAL206105-1G
SIAL206105-5G
Diammonium tetrachloroplatinate, Sigma-Aldrich®
Diammonium tetrachloroplatinate
Vzorec:
H₈Cl₄N₂Pt MW: 372.97 Bod tání: 140 Hustota: 2.93 g/cm³ (20 °C) |
MDL Number:
MFCD00010885 CAS číslo: 13820-41-2 EINECS: 237-499-1 UN: 3288 ADR: 6.1 III Merck Index: 12,00583 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...