Tisk…

Dipotassium tetrachloropalladate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Potassium tetrachloropalladate (II), Potassium chloropalladite

M-Clarity™ quality level = MQ200

205796-10G 205796-2G
SIAL205796-10GEA 25160 CZK
SIAL205796-10G SIAL205796-2G
Dipotassium tetrachloropalladate, Sigma-Aldrich®
Dipotassium tetrachloropalladate
Vzorec: Cl₄K₂Pd
MW: 326,43 g/mol
Bod tání: 105 °C
Hustota: 2,67 g/cm³ (20 °C)
MDL Number: MFCD00011373
CAS číslo: 10025-98-6
EINECS: 233-049-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR