2-(3-Hydroxyphenyl)ethanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Hydroxyphenethyl alcohol
M-Clarity™ quality level = MQ100
199028-10G
199028-1G
SIAL199028-10GEA
13600
CZK
SIAL199028-10G
SIAL199028-1G
2-(3-Hydroxyphenyl)ethanol, Sigma-Aldrich®
2-(3-Hydroxyphenyl)ethanol
Vzorec:
C₈H₁₀O₂ MW: 138,17 g/mol Bod varu: 124…126 °C (0,2 mmHg) Hustota: 1,082 g/cm³ (20 °C) |
CAS číslo:
13398-94-2 EINECS: 236-485-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...