Thiophene-3-carbaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde
M-Clarity™ quality level = MQ100
196282-10G
196282-1G
196282-5G
SIAL196282-10GEA
6830
CZK
SIAL196282-10G
SIAL196282-1G
SIAL196282-5G
Thiophene-3-carbaldehyde, Sigma-Aldrich®
Thiophene-3-carbaldehyde
Vzorec:
C₅H₄OS MW: 112,15 g/mol Bod varu: 191 °C (1013 hPa) Bod tání: –30 °C Hustota: 1,28 g/cm³ (24 °C) |
MDL Number:
MFCD00005466 CAS číslo: 498-62-4 EINECS: 207-865-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...