Iodotrimethylsilane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Trimethyliodosilane, Trimethylsilyl iodide
M-Clarity™ quality level = MQ200
195529-100G
195529-25G
195529-5G
SIAL195529-100GEA
12970
CZK
SIAL195529-100G
SIAL195529-25G
SIAL195529-5G
Iodotrimethylsilane, Sigma-Aldrich®
Iodotrimethylsilane
Vzorec:
C₃H₉ISi MW: 200.09 Bod varu: 106 Bod tání: < 0 °C Hustota: 1.47 g/cm³ (20 °C) |
MDL Number:
MFCD00001028 CAS číslo: 16029-98-4 EINECS: 240-171-0 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...