Acrolein dimethyl acetal, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
190543-25ML
190543-5ML
SIAL190543-25MLEA
9900
CZK
SIAL190543-25ML
SIAL190543-5ML
Acrolein dimethyl acetal, Sigma-Aldrich®
Acrolein dimethyl acetal
Vzorec:
C₅H₁₀O₂ Bod varu: 89 °C (1013 hPa) Hustota: 0,862 g/cm³ (20 °C) |
MDL Number:
MFCD00008632 CAS číslo: 6044-68-4 EINECS: 227-936-4 UN: 3384 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...