2-Amino-6-nitrobenzothiazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Nitrobenzothiazol-2-ylamine, 6-Nitro-1,3-benzothiazol-2-amine, 6-Nitrobenzo[d]thiazol-2-amine
M-Clarity™ quality level = MQ100
190241-100G
190241-25G
SIAL190241-100GEA
2830
CZK
SIAL190241-100G
SIAL190241-25G
2-Amino-6-nitrobenzothiazole, Sigma-Aldrich®
2-Amino-6-nitrobenzothiazole
Vzorec:
C₇H₅N₃O₂S MW: 195,2 g/mol Bod tání: 252 °C Hustota: 1,649 g/cm³ (20 °C) |
MDL Number:
MFCD00005786 CAS číslo: 6285-57-0 EINECS: 228-513-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...