Tisk…

2-(4-Chlorophenyl)ethanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Chlorophenethyl alcohol

M-Clarity™ quality level = MQ100

183423-10G 183423-50G
SIAL183423-10GEA 2430 CZK
SIAL183423-10G SIAL183423-50G
2-(4-Chlorophenyl)ethanol, Sigma-Aldrich®
2-(4-Chlorophenyl)ethanol
Vzorec: C₈H₉ClO
MW: 156,61 g/mol
Bod varu: 259 °C (1013 hPa)
Hustota: 1,189 g/cm³ (25 °C)
MDL Number: MFCD00002899
CAS číslo: 1875-88-3
EINECS: 217-506-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR