2-Amino-6-methoxybenzothiazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Methoxybenzothiazol-2-ylamine
M-Clarity™ quality level = MQ200
162590-250G
162590-50G
SIAL162590-250GEA
0
CZK
SIAL162590-250G
SIAL162590-50G
2-Amino-6-methoxybenzothiazole, Sigma-Aldrich®
2-Amino-6-methoxybenzothiazole
Vzorec:
C₈H₈N₂OS MW: 180,23 g/mol Bod varu: 348 °C (1013 hPa) Bod tání: 160 °C |
MDL Number:
MFCD00005787 CAS číslo: 1747-60-0 EINECS: 217-130-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...