4-Benzyloxyphenol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Monobenzone4-Hydroxyphenyl benzyl etherHydroquinone monobenzyl ether4-(Benzyloxy)phenol
M-Clarity™ quality level = MQ100
158348-100G
158348-25G
SIAL158348-100GEA
3400
CZK
SIAL158348-100G
SIAL158348-25G
4-Benzyloxyphenol, Sigma-Aldrich®
4-Benzyloxyphenol
Vzorec:
C₁₃H₁₂O₂ MW: 200,24 g/mol Bod varu: 328 °C (1013 hPa) Bod tání: 117,9…121.5 °C Hustota: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00002333 CAS číslo: 103-16-2 EINECS: 203-083-3 Merck Index: 12,06331 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...